Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003577.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003577.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003577.mm.1 |
TC1700000419.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003576.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003576.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003576.mm.1 |
TC1700000419.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003571.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003571.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003571.mm.1 |
TC1700000419.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003573.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003573.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003573.mm.1 |
TC1700000419.mm.1 |
MTA TC |