Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003582.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003582.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003582.mm.1 |
TC1700000419.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003580.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003580.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003580.mm.1 |
TC1700000419.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003584.mm.1 |
NM_029870.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003584.mm.1 |
NM_029870.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003584.mm.1 |
TC1700000419.mm.1 |
MTA TC |