Assay Details
| Gene Symbol: Trmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028063.2 | NP_082339.1 | 6 - 7 | 756 | 120 | |
| GenBank mRNA | AK005541.1 | - | 6 - 7 | 756 | 120 | |
| AK043532.1 | - | 6 - 7 | 763 | 120 | ||
| AK044059.1 | - | 6 - 7 | 763 | 120 | ||
| AY349617.1 | - | 6 - 7 | 763 | 120 | ||
| BC051548.1 | - | 6 - 7 | 757 | 120 | ||
| BC061026.1 | - | 6 - 7 | 743 | 30305575 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmu
Genomic Map