Assay Details
| Gene Symbol: Gtse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168672.1 | NP_001162143.1 | 3 - 4 | 239 | 138 | |
| NM_013882.2 | NP_038910.1 | 2 - 3 | 209 | 138 | ||
| GenBank mRNA | AJ222580.1 | - | 2 - 3 | 179 | 138 | |
| AK012870.1 | - | 3 - 4 | 240 | 138 | ||
| AK081582.1 | - | 3 - 4 | 202 | 138 | ||
| BC027213.1 | - | 2 - 3 | 210 | 4039841 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ222580.1, AK012870.1, AK081582.1, BC027213.1, NM_001168672.1, NM_013882.2 transcripts.
Set Membership: |
|

Gtse1
Genomic Map