Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500006492.mm.1 |
NM_001168672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006492.mm.1 |
NM_001168672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006492.mm.1 |
NM_013882.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006492.mm.1 |
NM_013882.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006492.mm.1 |
TC1500000826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500006493.mm.1 |
NM_001168672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006493.mm.1 |
NM_001168672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006493.mm.1 |
NM_013882.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006493.mm.1 |
NM_013882.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500006493.mm.1 |
TC1500000826.mm.1 |
MTA TC |