Assay Details
| Gene Symbol: Slc26a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290320.1 | NP_001277249.1 | 9 - 10 | 1447 | 74 | |
| GenBank mRNA | BC027076.1 | - | 9 - 10 | 1447 | 5361175 | 74 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc26a8
Genomic Map