Assay Details
| Gene Symbol: Scube3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004366.1 | NP_001004366.1 | 5 - 6 | 599 | 64 | |
| GenBank mRNA | AY639609.1 | - | 5 - 6 | 599 | 64 | |
| BC079849.1 | - | 3 - 4 | 348 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scube3
Genomic Map