Assay Details
| Gene Symbol: Scube3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004366.1 | NP_001004366.1 | 8 - 9 | 949 | 62 | |
| GenBank mRNA | AK083515.1 | - | 2 - 3 | 288 | 62 | |
| AY639609.1 | - | 8 - 9 | 949 | 62 | ||
| BC079849.1 | - | 6 - 7 | 698 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scube3
Genomic Map