Assay Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 8 - 9 | 1815 | 105 | |
| GenBank mRNA | AK033354.1 | - | 8 - 9 | 1815 | 105 | |
| AK075652.1 | - | 1 - 2 | 594 | 105 | ||
| BC070460.1 | - | 8 - 9 | 1188 | 30548562 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srek1
Genomic Map