Assay Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 2 - 3 | 868 | 143 | |
| GenBank mRNA | AK033354.1 | - | 2 - 3 | 868 | 143 | |
| AK037115.1 | - | 2 - 3 | 844 | 143 | ||
| AK039311.1 | - | 4 - 5 | 603 | 143 | ||
| AK044086.1 | - | 1 - 2 | 172 | 143 | ||
| AK052265.1 | - | 4 - 5 | 737 | 143 | ||
| AK054414.1 | - | 2 - 3 | 1319 | 143 | ||
| BC063761.1 | - | 5 - 6 | 613 | 143 | ||
| BC070460.1 | - | 2 - 3 | 241 | 30548562 | 143 | |
More Information
Set Membership: |
|

Srek1
Genomic Map