Assay Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 3 - 4 | 1043 | 96 | |
| GenBank mRNA | AK033354.1 | - | 3 - 4 | 1043 | 96 | |
| AK037115.1 | - | 3 - 4 | 1019 | 96 | ||
| AK039311.1 | - | 5 - 6 | 778 | 96 | ||
| AK044086.1 | - | 2 - 3 | 347 | 96 | ||
| AK052265.1 | - | 5 - 6 | 912 | 96 | ||
| AK054414.1 | - | 3 - 4 | 1494 | 96 | ||
| BC063761.1 | - | 6 - 7 | 788 | 96 | ||
| BC070460.1 | - | 3 - 4 | 416 | 30548562 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srek1
Genomic Map