Assay Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 5 - 6 | 1295 | 84 | |
| GenBank mRNA | AK028794.1 | - | 1 - 2 | 2831 | 84 | |
| AK033354.1 | - | 5 - 6 | 1295 | 84 | ||
| AK037115.1 | - | 5 - 6 | 1271 | 84 | ||
| AK039311.1 | - | 7 - 8 | 1030 | 84 | ||
| AK044086.1 | - | 4 - 5 | 599 | 84 | ||
| AK052265.1 | - | 7 - 8 | 1164 | 84 | ||
| AK054414.1 | - | 5 - 6 | 1934 | 84 | ||
| BC063761.1 | - | 8 - 9 | 1040 | 84 | ||
| BC070460.1 | - | 5 - 6 | 668 | 30548562 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srek1
Genomic Map