Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_001093759.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_001093759.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_001093760.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_001093760.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_025879.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
NM_025879.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017668.mm.1 |
TC1300002596.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_001093759.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_001093759.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_001093760.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_001093760.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_025879.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
NM_025879.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300017670.mm.1 |
TC1300002596.mm.1 |
MTA TC |