Assay Details
| Gene Symbol: Dnajc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030046.2 | NP_084322.2 | 6 - 7 | 1051 | 85 | |
| GenBank mRNA | AK019636.1 | - | 1 - 2 | 146 | 85 | |
| AK141303.1 | - | 3 - 4 | 511 | 85 | ||
| BC048354.1 | - | 2 - 3 | 342 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc21
Genomic Map