Assay Details
| Gene Symbol: Dnajc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030046.2 | NP_084322.2 | 8 - 9 | 1292 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc21| Gene Symbol: Dnajc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030046.2 | NP_084322.2 | 8 - 9 | 1292 | 138 | |
Set Membership: |
|