Assay Details
| Gene Symbol: Scube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271472.1 | NP_001258401.1 | 11 - 12 | 1429 | 65 | |
| NM_001271473.1 | NP_001258402.1 | 8 - 9 | 1096 | 65 | ||
| NM_022723.3 | NP_073560.2 | 10 - 11 | 1339 | 65 | ||
| GenBank mRNA | AF276425.1 | - | 10 - 11 | 1206 | 65 | |
| AK041590.1 | - | 10 - 11 | 1322 | 65 | ||
| BC066066.1 | - | 11 - 12 | 1387 | 6850999 | 65 | |
| BC080278.1 | - | 8 - 9 | 1064 | 30354974 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scube1
Genomic Map