Assay Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146027.2 | NP_666139.1 | 2 - 3 | 278 | 73 | |
| GenBank mRNA | AK075869.1 | - | 2 - 3 | 242 | 73 | |
| BC010784.1 | - | 2 - 3 | 229 | 73 | ||
| BC021346.1 | - | 2 - 3 | 247 | 4163541 | 73 | |
| CN691874.1 | - | 2 - 3 | 278 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scrn2
Genomic Map