Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_001199025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_016915.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
NM_016915.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014589.mm.1 |
TC1500001868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_001199025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_016915.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
NM_016915.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500014590.mm.1 |
TC1500001868.mm.1 |
MTA TC |