Assay Details
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207225.1 | NP_997108.1 | 2 - 3 | 210 | 58 | |
GenBank mRNA | AK155250.1 | - | 3 - 4 | 396 | 58 | |
BC066052.1 | - | 2 - 3 | 210 | 6827645 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
