Assay Details
| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134252.3 | NP_599013.1 | 12 - 13 | 74 | ||
| GenBank mRNA | AF481480.2 | - | 1808 | 74 | ||
| AY095352.1 | - | 10 - 11 | 74 | |||
| BC117934.1 | - | 12 - 13 | 1539 | 40097778 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpm8
Genomic Map