Assay Details
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134252.3 | NP_599013.1 | 23 - 24 | 3312 | 126 | |
GenBank mRNA | AF481480.2 | - | 3581 | 126 | ||
AY095352.1 | - | 21 - 22 | 3146 | 126 | ||
BC117934.1 | - | 23 - 24 | 3312 | 40097778 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
