Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014356.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014356.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014356.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014355.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014355.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014355.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014357.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014357.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014357.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014358.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014358.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014358.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014359.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014359.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014359.mm.1 |
TC1700001624.mm.1 |
MTA TC |