Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014348.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014348.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014348.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014347.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014347.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014347.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014349.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014349.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014349.mm.1 |
TC1700001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014350.mm.1 |
NM_001031621.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014350.mm.1 |
NM_001031621.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014350.mm.1 |
TC1700001624.mm.1 |
MTA TC |