Assay Details
Gene Symbol: Abca17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031621.2 | NP_001026792.2 | 5 - 6 | 863 | 102 | |
GenBank mRNA | AB112584.1 | - | 5 - 6 | 863 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
