Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500005882.mm.1 |
NM_011889.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005882.mm.1 |
NM_011889.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005882.mm.1 |
TC1500000747.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005878.mm.1 |
NM_011889.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005878.mm.1 |
NM_011889.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005878.mm.1 |
TC1500000747.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005879.mm.1 |
NM_011889.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005879.mm.1 |
NM_011889.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005879.mm.1 |
TC1500000747.mm.1 |
MTA TC |