Assay Details
| Gene Symbol: Slc9a3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023055.2 | NP_075542.2 | 1 - 2 | 366 | 66 | |
| GenBank mRNA | AB026489.1 | - | 1 - 2 | 339 | 66 | |
| AK004710.1 | - | 1 - 2 | 360 | 66 | ||
| AK039491.1 | - | 1 - 2 | 354 | 66 | ||
| AK075813.1 | - | 1 - 2 | 367 | 66 | ||
| AK084801.1 | - | 1 - 2 | 367 | 66 | ||
| AK169007.1 | - | 1 - 2 | 356 | 66 | ||
| AK170057.1 | - | 1 - 2 | 352 | 66 | ||
| BC070458.1 | - | 1 - 2 | 350 | 30603578 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r2
Genomic Map