Assay Details
| Gene Symbol: Slc9a3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023055.2 | NP_075542.2 | 3 - 4 | 753 | 110 | |
| NM_023449.3 | NP_075938.2 | 2 - 3 | 323 | 110 | ||
| GenBank mRNA | AB026489.1 | - | 3 - 4 | 726 | 110 | |
| AK002557.1 | - | 2 - 3 | 324 | 110 | ||
| AK039491.1 | - | 3 - 4 | 741 | 110 | ||
| AK075800.1 | - | 3 - 4 | 756 | 110 | ||
| AK075813.1 | - | 3 - 4 | 754 | 110 | ||
| AK084801.1 | - | 3 - 4 | 754 | 110 | ||
| AK169007.1 | - | 3 - 4 | 743 | 110 | ||
| AK170057.1 | - | 3 - 4 | 739 | 110 | ||
| BC065778.1 | - | 2 - 3 | 294 | 3467172 | 110 | |
| BC070458.1 | - | 3 - 4 | 737 | 30603578 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r2
Genomic Map