Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014184.mm.1 |
NM_001080773.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014184.mm.1 |
NM_001286662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014184.mm.1 |
NM_011062.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014184.mm.1 |
NM_011062.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014184.mm.1 |
TC1700001615.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014187.mm.1 |
NM_001080773.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014187.mm.1 |
NM_001286662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014187.mm.1 |
NM_011062.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014187.mm.1 |
NM_011062.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014187.mm.1 |
TC1700001615.mm.1 |
MTA TC |