Assay Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027937.2 | NP_082213.2 | 13 - 14 | 1514 | 89 | |
| GenBank mRNA | AK129327.1 | - | 13 - 14 | 1290 | 89 | |
| BC059916.1 | - | 4 - 5 | 346 | 89 | ||
| BC060720.1 | - | 13 - 14 | 1278 | 89 | ||
| BC138443.1 | - | 13 - 14 | 1422 | 8861463 | 89 | |
| BC145281.1 | - | 13 - 14 | 1422 | 9053835 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin1
Genomic Map