| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_001024716.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_001024716.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_001039156.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_001039156.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_138579.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
NM_138579.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004899.mm.1 |
TC1500000660.mm.1 |
MTA TC |