Assay Details
| Gene Symbol: Sox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011437.1 | NP_035567.1 | 1 - 2 | 214 | 131 | |
| GenBank mRNA | AF017182.1 | - | 2 - 3 | 276 | 131 | |
| AK042542.1 | - | 1 - 2 | 215 | 131 | ||
| AK043220.1 | - | 1 - 2 | 211 | 131 | ||
| BC023356.1 | - | 1 - 2 | 48 | 5027211 | 131 | |
| BC025171.1 | - | 1 - 2 | 177 | 4912940 | 131 | |
| BC051058.1 | - | 1 - 2 | 48 | 131 | ||
| U66141.1 | - | 1 - 2 | 212 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK043220.1, BC023356.1, BC025171.1, NM_011437.1 transcripts.
Set Membership: |
|

Sox10
Genomic Map