Assay Details
| Gene Symbol: Eif3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145139.2 | NP_660121.2 | 10 - 11 | 1118 | 108 | |
| GenBank mRNA | AJ310346.1 | - | 10 - 11 | 1128 | 108 | |
| AK032936.1 | - | 10 - 11 | 1106 | 108 | ||
| AK075584.1 | - | 10 - 11 | 1124 | 108 | ||
| AK150895.1 | - | 10 - 11 | 1113 | 108 | ||
| AK151175.1 | - | 10 - 11 | 1113 | 108 | ||
| AK167640.1 | - | 10 - 11 | 1113 | 108 | ||
| AK168009.1 | - | 10 - 11 | 1116 | 108 | ||
| AK168295.1 | - | 10 - 11 | 1115 | 108 | ||
| BC021351.1 | - | 1786 | 108 | |||
| BC024463.1 | - | 10 - 11 | 1118 | 4975621 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3l
Genomic Map