Assay Details
| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027231.1 | NP_081507.1 | 4 - 5 | 340 | 75 | |
| GenBank mRNA | AK007913.1 | - | 4 - 5 | 355 | 75 | |
| AK165600.1 | - | 4 - 5 | 367 | 75 | ||
| BC024419.1 | - | 4 - 5 | 340 | 4983359 | 75 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364608 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364609 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292959 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103013 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111345 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P276222 | AK169434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P276222 | BC024419.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P276222 | NM_027231.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P276222 | AK007913.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P276222 | AK165600.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30364609 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103013 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111345 | - | - |
| Whole Mouse Genome G4122A | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | AK169434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | BC024419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | NM_027231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | AK007913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P120729 | AK165600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276222 | AK169434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276222 | BC024419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276222 | NM_027231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276222 | AK007913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276222 | AK165600.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111345 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12011184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12011178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364608 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30364738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12091945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12392174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12050493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12409116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12252204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12123052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12242565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12422329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11842358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12409122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12377331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12242560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11972126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12378805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12145780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12378838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364704 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425262 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425258 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415754_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427402_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425257 | - | - |
| Mouse Genome 430 2.0 Array | 1415754_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427402_at | - | - |
| Mouse Genome 430 2.0 Array | 1427402_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415754_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69833_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69833_at | - | - |
| Mouse Expression Set 430 | 1415754_PM_at | - | - |
| Mouse Expression Set 430 | 1415754_at | - | - |
| Mouse Expression Set 430 | 1415754_at | - | - |
| Mouse Expression Set 430 | 1415754_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415754_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415754_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415754_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415754_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427402_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427402_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415754_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415754_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 706194 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237750 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237750 | - | - |

Polr2f
Genomic Map