| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253817.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253817.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_001253820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014632.mm.1 |
TC1500001870.mm.1 |
MTA TC |