Assay Details
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010668.2 | NP_034798.2 | 3 - 4 | 999 | 68 | |
GenBank mRNA | AK132476.1 | - | 3 - 4 | 1001 | 68 | |
AK161078.1 | - | 3 - 4 | 998 | 68 | ||
AK161098.1 | - | 3 - 4 | 1001 | 68 | ||
BC120485.1 | - | 3 - 4 | 965 | 40129408 | 68 | |
BC139485.1 | - | 3 - 4 | 965 | 8860587 | 68 | |
X74784.1 | - | 3 - 4 | 965 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
