Assay Details
| Gene Symbol: Acvr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007395.3 | NP_031421.1 | 6 - 7 | 1161 | 112 | |
| GenBank mRNA | AK154578.1 | - | 1184 | 112 | ||
| AK157910.1 | - | 6 - 7 | 1162 | 112 | ||
| BC059832.1 | - | 1154 | 112 | |||
| BC145775.1 | - | 6 - 7 | 1214 | 40131078 | 112 | |
| BC145777.1 | - | 6 - 7 | 1214 | 40131080 | 112 | |
| Z31663.1 | - | 6 - 7 | 1150 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr1b
Genomic Map