Assay Details
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010444.2 | NP_034574.1 | 1 - 2 | 122 | 65 | |
GenBank mRNA | AK004709.1 | - | 1 - 2 | 116 | 65 | |
AK157203.1 | - | 1 - 2 | 122 | 65 | ||
BC004770.1 | - | 1 - 2 | 102 | 3156434 | 65 | |
J04113.1 | - | 1 - 2 | 105 | 65 | ||
X60132.1 | - | 1 - 2 | 72 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004709.1, AK157203.1, BC004770.1, J04113.1, NM_010444.2, X60132.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
