Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 17 | 120 | ||
| NM_011815.5 | NP_035945.1 | 18 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 17 | 120 | ||
| NM_011815.5 | NP_035945.1 | 18 | 120 | |||
Set Membership: |
|