Assay Details
Gene Symbol: Ddx23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080981.1 | NP_001074450.1 | 6 - 7 | 683 | 79 | |
GenBank mRNA | AK014820.1 | - | 4 - 5 | 1441 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
