Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500017966.mm.1 |
NM_029098.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017966.mm.1 |
NM_029098.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017966.mm.1 |
TC1500002197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017971.mm.1 |
NM_029098.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017971.mm.1 |
NM_029098.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017971.mm.1 |
TC1500002197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017965.mm.1 |
NM_029098.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017965.mm.1 |
NM_029098.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017965.mm.1 |
TC1500002197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017969.mm.1 |
NM_029098.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017969.mm.1 |
NM_029098.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017969.mm.1 |
TC1500002197.mm.1 |
MTA TC |