Assay Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038658.2 | NP_001033747.1 | 6 - 7 | 1026 | 62 | |
| NM_028224.4 | NP_082500.2 | 6 - 7 | 1062 | 62 | ||
| GenBank mRNA | AF468028.1 | - | 5 - 6 | 491 | 62 | |
| AK013476.1 | - | 6 - 7 | 465 | 62 | ||
| AK030513.1 | - | 6 - 7 | 549 | 62 | ||
| AK080839.1 | - | 6 - 7 | 576 | 62 | ||
| AK129246.1 | - | 6 - 7 | 575 | 62 | ||
| AK136791.1 | - | 6 - 7 | 544 | 62 | ||
| AK158960.1 | - | 6 - 7 | 855 | 62 | ||
| BC072560.1 | - | 6 - 7 | 1062 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faim2
Genomic Map