Assay Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038658.2 | NP_001033747.1 | 7 - 8 | 1069 | 101 | |
| NM_028224.4 | NP_082500.2 | 7 - 8 | 1105 | 101 | ||
| GenBank mRNA | AK013476.1 | - | 7 - 8 | 508 | 101 | |
| AK030513.1 | - | 7 - 8 | 592 | 101 | ||
| AK080839.1 | - | 7 - 8 | 619 | 101 | ||
| AK129246.1 | - | 7 - 8 | 618 | 101 | ||
| AK136791.1 | - | 7 - 8 | 587 | 101 | ||
| AK158960.1 | - | 7 - 8 | 898 | 101 | ||
| BC072560.1 | - | 7 - 8 | 1105 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faim2
Genomic Map