Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500017930.mm.1 |
NM_026967.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017930.mm.1 |
NM_026967.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017930.mm.1 |
TC1500002194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017931.mm.1 |
NM_026967.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017931.mm.1 |
NM_026967.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017931.mm.1 |
TC1500002194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017926.mm.1 |
NM_026967.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017926.mm.1 |
NM_026967.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017926.mm.1 |
TC1500002194.mm.1 |
MTA TC |