Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700016107.mm.1 |
NM_001085555.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016107.mm.1 |
NM_001085555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016107.mm.1 |
NM_013687.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016107.mm.1 |
NM_013687.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016107.mm.1 |
TC1700001750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016108.mm.1 |
NM_001085555.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016108.mm.1 |
NM_001085555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016108.mm.1 |
NM_013687.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016108.mm.1 |
NM_013687.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016108.mm.1 |
TC1700001750.mm.1 |
MTA TC |