Assay Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029624.4 | NP_083900.3 | 2 - 3 | 511 | 64 | |
| GenBank mRNA | AK010312.1 | - | 2 - 3 | 482 | 64 | |
| AK053241.1 | - | 2 - 3 | 488 | 64 | ||
| AK154622.1 | - | 2 - 3 | 508 | 64 | ||
| AK169983.1 | - | 2 - 3 | 509 | 64 | ||
| AK172374.1 | - | 2 - 3 | 490 | 64 | ||
| BC020104.1 | - | 2 - 3 | 518 | 3963699 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf1
Genomic Map