Assay Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018857.1 | NP_061345.1 | 3 - 4 | 287 | 77 | |
| GenBank mRNA | AK144391.1 | - | 4 - 5 | 219 | 77 | |
| AK146799.1 | - | 3 - 4 | 337 | 77 | ||
| BC023753.1 | - | 3 - 4 | 291 | 5354484 | 77 | |
| D86370.1 | - | 3 - 4 | 287 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msln
Genomic Map