Assay Details
| Gene Symbol: Rhbdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144816.1 | NP_659065.1 | 7 - 8 | 951 | 62 | |
| GenBank mRNA | AK140428.1 | - | 7 - 8 | 982 | 62 | |
| BC021549.1 | - | 7 - 8 | 951 | 4501997 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106621 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106622 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628426 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110132 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106631 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110132 | - | - |
| Whole Mouse Genome G4122A | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256757 | AK140428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256757 | BC021549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256757 | NM_144816.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106318 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110132 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13139174 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13139165 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13403483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13139176 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106621 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31106620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13403530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13403510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13403495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449065 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426094_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449061 | - | - |
| Mouse Genome 430 2.0 Array | 1426094_at | - | - |
| Mouse Genome 430 2.0 Array | 1426094_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214951_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214951_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700015343.mm.1 | NM_144816.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015343.mm.1 | NM_144816.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015343.mm.1 | TC1700002801.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700015344.mm.1 | NM_144816.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015344.mm.1 | NM_144816.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015344.mm.1 | TC1700002801.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426094_at | - | - |
| Mouse Expression Set 430 | 1426094_PM_at | - | - |
| Murine Genome U74v2 Set | 138548_at | - | - |
| Murine Genome U74v2 Set | 138548_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426094_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426094_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426094_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 484616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1500736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2710802 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2710802 | - | - |
| mouse-6 v1.1 (Illumina) | scl50168.8.1_112-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50168.8.1_112-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2710802 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2710802 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50168.8.1_112-S | - | - |

Rhbdl1
Genomic Map