Assay Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163661.1 | NP_001157133.1 | 7 - 8 | 1568 | 134 | |
| NM_027880.2 | NP_082156.2 | 7 - 8 | 1479 | 134 | ||
| GenBank mRNA | AK004582.1 | - | 7 - 8 | 1349 | 134 | |
| AK028658.1 | - | 7 - 8 | 1333 | 134 | ||
| AK050773.1 | - | 7 - 8 | 1298 | 134 | ||
| AK084830.1 | - | 7 - 8 | 1349 | 134 | ||
| AK129189.1 | - | 7 - 8 | 1561 | 134 | ||
| AK143827.1 | - | 7 - 8 | 1569 | 134 | ||
| AK146963.1 | - | 7 - 8 | 1355 | 134 | ||
| AK154727.1 | - | 7 - 8 | 1381 | 134 | ||
| BC011077.1 | - | 7 - 8 | 1329 | 3984911 | 134 | |
| BC080713.1 | - | 7 - 8 | 1508 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Telo2
Genomic Map