Assay Details
| Gene Symbol: 1700074H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045296.1 | 2 - 3 | 283 | 129 | ||
| GenBank mRNA | AK006947.1 | - | 2 - 3 | 294 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700074H08Rik| Gene Symbol: 1700074H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045296.1 | 2 - 3 | 283 | 129 | ||
| GenBank mRNA | AK006947.1 | - | 2 - 3 | 294 | 129 | |
Set Membership: |
|