Assay Details
| Gene Symbol: 5031439G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033273.2 | NP_001028445.1 | 2 - 3 | 515 | 79 | |
| GenBank mRNA | AK138575.1 | - | 1 - 2 | 500 | 79 | |
| AK149782.1 | - | 2 - 3 | 516 | 79 | ||
| AK170532.1 | - | 2 - 3 | 548 | 79 | ||
| AK173065.1 | - | 2 - 3 | 273 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

5031439G07Rik
Genomic Map